Learn More
Biomatik Corporation Mouse Bone morphogenetic protein 1(BMP1) ELISA kit
Supplier:  Biomatik Corporation EKC42383
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.81 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P98063. Target Synonyms: FLJ44432, PCOLC, PCP, TLD, pCP-2, procollagen C-endopeptidase|procollagen C-proteinase. Restriction: For Research Use Only.
Catalog No. 50-229-5067