Learn More
Biomatik Corporation Human Lamin A/C (Lmna) Elisa Kit
Supplier:  Biomatik Corporation EKC34365
Encompass_Preferred
Human Lamin A/C (Lmna) Elisa Kit for detecting Lamin A/C in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02545; LMNA. Target Synonyms: 70 kDa lamin; Cardiomyopathy dilated 1A (autosomal dominant); CDCD1; CDDC; CMD1A; CMT2B1; EMD2; FPL; FPLD; FPLD2; HGPS; IDC; Lamin A; Lamin A/C; Lamin A/C like 1; Lamin; Lamin C; lamin-a; Lamin-A/C; LDP1; LFP; LGMD1B; Limb girdle muscular dystrophy 1B (autosomal dominant); LMN 1; LMN A; LMN C; LMN1; LMNA; LMNA_HUMAN; LMNC; LMNL1; Prelamin A/C; PRO1; Renal carcinoma antigen NY REN 32; Renal carcinoma antigen NY-REN-32; Renal carcinoma antigen NYREN32. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4087