Learn More
Biomatik Corporation Human Layilin (Layn) Elisa Kit
Supplier:  Biomatik Corporation EKC34381
Encompass_Preferred
Human Layilin (Layn) Elisa Kit for detecting Layilin in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q6UX15; LAYN. Target Synonyms: Layilin; LAYN; LAYN_HUMAN; UNQ208/PRO234. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4091