Learn More
Biomatik Corporation Human Low-Density Lipoprotein-Receptor-Related Protein 1, Lrp-1 Elisa Kit
Supplier:  Biomatik Corporation EKC34473
Encompass_Preferred
Human Low-Density Lipoprotein-Receptor-Related Protein 1, Lrp-1 Elisa Kit for detecting Low Density Lipoprotein-Related Protein 1 (Alpha-2-Macroglobulin Receptor) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q07954; LRP1. Target Synonyms: A2MR; Alpha 2 macroglobulin receptor; alpha 2MR; Alpha-2-macroglobulin receptor; APOER; Apolipoprotein E receptor; APR; CD 91; CD91; CD91 antigen; IGFBP3R; LDL receptor related protein 1; Low density lipoprotein receptor related protein 1; Low density lipoprotein related protein 1; Low-density lipoprotein receptor-related protein 1 intracellular domain; LRP 1; LRP 515; LRP 85; LRP; LRP ICD; LRP-1; LRP-515; LRP-85; Lrp1; LRP1 protein; LRP1_HUMAN; LRP1A; LRP515; LRP85; LRPICD. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4106