Learn More
Biomatik Corporation Human Ly6/Plaur Domain-Containing Protein 1 (Lypd1) Elisa Kit
Supplier:  Biomatik Corporation EKC34482
Encompass_Preferred
Human Ly6/Plaur Domain-Containing Protein 1 (Lypd1) Elisa Kit for detecting Ly6/Plaur Domain Containing 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8N2G4; LYPD1. Target Synonyms: FLJ41033; LY6/PLAUR domain containing 1; Ly6/PLAUR domain-containing protein 1; Lypd1; LYPD1_HUMAN; LYPDC1; MGC29643; PHTS; Putative HeLa tumor suppressor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4108