Learn More
Biomatik Corporation Human Lysyl Oxidase Homolog 3 (Loxl3) Elisa Kit
Supplier:  Biomatik Corporation EKC34511
Encompass_Preferred
Human Lysyl Oxidase Homolog 3 (Loxl3) Elisa Kit for detecting Lysyl Oxidase Homolog 3(Loxl3) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P58215; LOXL3. Target Synonyms: LOXL; Loxl3; LOXL3_HUMAN; Lysyl oxidase homolog 3; Lysyl oxidase like 3; Lysyl oxidase-like protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4114