Learn More
Biomatik Corporation Human Macroprolactin Elisa Kit
Supplier:  Biomatik Corporation EKC34522
Encompass_Preferred
Human Macroprolactin Elisa Kit for detecting Macroprolactin in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.4-20uIU/ml. Sensitivity: 0.5uIU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4118