Learn More
Biomatik Corporation Human Melanin-Concentrating Hormone (Pmch) Elisa Kit
Supplier:  Biomatik Corporation EKC34571
Encompass_Preferred
Human Melanin-Concentrating Hormone (Pmch) Elisa Kit for detecting Melanin-Concentrating Hormone (Pmch) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P20382; PMCH. Target Synonyms: PMCH; MCHPro-MCH [Cleaved into: Neuropeptide-glycine-glutamic acid; NGE; Neuropeptide G-E); Neuropeptide-glutamic acid-isoleucine; NEI; Neuropeptide E-I); Melanin-concentrating hormone; MCH)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4126