Learn More
Biomatik Corporation Human Membrane-Associated Progesterone Receptor Component 1 (Pgrmc1) Elisa Kit
Supplier:  Biomatik Corporation EKC34589
Encompass_Preferred
Human Membrane-Associated Progesterone Receptor Component 1 (Pgrmc1) Elisa Kit for detecting Progesterone Receptor Membrane Component 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 47-3000pg/ml. Sensitivity: 11.7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O00264; PGRMC1. Target Synonyms: PGRMC1; HPR6.6; PGRMC; Membrane-associated progesterone receptor component 1; mPR; Dap1; IZA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4130