Learn More
Biomatik Corporation Human Methylcrotonoyl-CoA carboxylase subunit alpha, mitochondrial(MCCC1) ELISA kit
Supplier:  Biomatik Corporation EKC34604
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78 ng/ml-50ng/ml. Sensitivity: 0.19 ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q96RQ3. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4135