Learn More
Biomatik Corporation Human Mitochondrial Import Inner Membrane Translocase Subunit Tim14 (Dnajc19) Elisa Kit
Supplier:  Biomatik Corporation EKC34625
Encompass_Preferred
Human Mitochondrial Import Inner Membrane Translocase Subunit Tim14 (Dnajc19) Elisa Kit for detecting Dnaj (Hsp40) Homolog, Subfamily C, Member 19 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 28-1800pg/ml. Sensitivity: 7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q96DA6; DNAJC19. Target Synonyms: DnaJ (Hsp40) homolog; subfamily C; member 19; DnaJ homolog subfamily C member 19; DNAJC19; homolog of yeast TIM14; Mitochondrial import inner membrane translocase subunit TIM14; PAM18; TIM 14; TIM14; TIM14_HUMAN; TIMM 14; TIMM14; translocase of the inner mitochondrial membrane 14; Translocase of the inner mitochondrial membrane 14; yeast homolog of. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4143