Learn More
Biomatik Corporation Human Myosin Light Chain Kinase, Mlck Elisa Kit
Supplier:  Biomatik Corporation EKC34707
Encompass_Preferred
Human Myosin Light Chain Kinase, Mlck Elisa Kit for detecting Myosin Light Chain Kinase in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q15746; MYLK. Target Synonyms: deglutamylated form; DKFZp686I10125; EC 2.7.11.18; FLJ12216; Kinase related protein; Kinase-related protein; KRP; MLCK; MLCK1; MLCK108; MLCK210; MSTP083; MYLK; MYLK_HUMAN; MYLK1; Myosin light chain kinase; Myosin light polypeptide kinase; OTTHUMP00000180642; OTTHUMP00000180643; smMLCK; smooth muscle; Smooth muscle myosin light chain kinase; Telokin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4154