Learn More
Biomatik Corporation Human Neuregulin-2 (Nrg2) Elisa Kit
Supplier:  Biomatik Corporation EKC34771
Encompass_Preferred
Human Neuregulin-2 (Nrg2) Elisa Kit for detecting Neuregulin-2 (Nrg2) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O14511; NRG2. Target Synonyms: Divergent of neuregulin 1; Divergent of neuregulin-1; Don 1; DON-1; Neural- and thymus-derived activator for ERBB kinases; Neuregulin 2; Neuregulin-2; NRG-2; Nrg2; NRG2_HUMAN; NTAK; Pro NRG2; Pro-NRG2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4164