Learn More
Biomatik Corporation Human Neurotrophin-3, Nt-3 Elisa Kit
Supplier:  Biomatik Corporation EKC34811
Encompass_Preferred
Human Neurotrophin-3, Nt-3 Elisa Kit for detecting Neurotrophin-3, Nt-3 in serum, plasma, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P20783; NTF3. Target Synonyms: HDNF; MGC129711; Nerve growth factor 2; Neurotrophic factor; Neurotrophin 3 precursor; Neurotrophin-3; Neurotrophin3; NGF 2; NGF-2; NGF2; NT 3; NT-3; NT3; NTF 3; NTF3; NTF3_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4169