Learn More
Biomatik Corporation Human N-Sulphoglucosamine Sulphohydrolase (Sgsh) Elisa Kit
Supplier:  Biomatik Corporation EKC34844
Encompass_Preferred
Human N-Sulphoglucosamine Sulphohydrolase (Sgsh) Elisa Kit for detecting N-Sulfoglucosamine Sulfohydrolase in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P51688; SGSH. Target Synonyms: Heparan sulfate sulfatase; Heparan sulphate sulphatase; HSS; MPS 3A; MPS3 A; MPS3A; Mucopolysaccharidosis type IIIA; N sulfoglucosamine sulfohydrolase (sulfamidase); N sulfoglucosamine sulfohydrolase; N-sulphoglucosamine sulphohydrolase; SFMD; SGSH; SPHM_HUMAN; Sulfoglucosamine sulfamidase; Sulphamidase; Sulphoglucosamine sulphamidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4176