Learn More
Biomatik Corporation Human Ornithine Carbamoyltransferase, Mitochondrial (Otc) Elisa Kit
Supplier:  Biomatik Corporation EKC34889
Encompass_Preferred
Human Ornithine Carbamoyltransferase, Mitochondrial (Otc) Elisa Kit for detecting Ornithine Carbamoyltransferase in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.25-80mU/ml. Sensitivity: 0.312mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P00480; OTC. Target Synonyms: EC 2.1.3.3; MGC129967; MGC129968; MGC138856; OCTD; Ornithine Carbamoyltransferase; Ornithine carbamoyltransferase mitochondrial; Ornithine carbamoyltransferase; mitochondrial; Ornithine transcarbamylase; Otc; OTC_HUMAN; OTCase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4192