Learn More
Biomatik Corporation Human Peptidyl-Prolyl Cis-Trans Isomerase C (Ppic) Elisa Kit
Supplier:  Biomatik Corporation EKC34949
Encompass_Preferred
Human Peptidyl-Prolyl Cis-Trans Isomerase C (Ppic) Elisa Kit for detecting Peptidylprolyl Isomerase C (Cyclophilin C) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P45877; PPIC. Target Synonyms: Cyclophilin C; CYPC; MGC3673; Parvulin; Peptidyl prolyl cis trans isomerase C; Peptidyl-prolyl cis-trans isomerase C; Peptidylprolyl isomerase C (cyclophilin C); peptidylprolyl isomerase C; PPIase C; ppiC; PPIC_HUMAN; Rotamase C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4203