Learn More
Biomatik Corporation Human Peroxiredoxin-6 (Prdx6) Elisa Kit
Supplier:  Biomatik Corporation EKC34966
Encompass_Preferred
Human Peroxiredoxin-6 (Prdx6) Elisa Kit for detecting Peroxiredoxin 6 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 12.5-800pg/ml. Sensitivity: 3.12pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P30041; PRDX6. Target Synonyms: 1 Cys; 1 Cys peroxiredoxin; 1 Cys PRX; 1 cysPrx; 1-Cys peroxiredoxin; 1-Cys PRX; 24 kDa protein; 9430088D19Rik; AA690119; Acidic calcium independent phospholipase A2; Acidic calcium-independent phospholipase A2; aiPLA2; Antioxidant protein 2; AOP2; Aop2 rs3; Brp 12; Ciliary body glutathione peroxidase; CP 3; EC 1.11.1.15; EC 1.11.1.7; EC 3.1.1.; Epididymis secretory sperm binding protein Li 128m; GPx; HEL S 128m; KIAA0106; Liver 2D page spot 40; Ltw4; Lvtw 4; MGC46173; mKIAA0106; Non selenium glutathione peroxidase; Non-selenium glutathione peroxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4207