Learn More
Biomatik Corporation Human Phospholipid Hydroperoxide Glutathione Peroxidase, Mitochondrial (Gpx4) Elisa Kit
Supplier:  Biomatik Corporation EKC35012
Encompass_Preferred
Human Phospholipid Hydroperoxide Glutathione Peroxidase, Mitochondrial (Gpx4) Elisa Kit for detecting Glutathione Peroxidase 4 (Phospholipid Hydroperoxidase) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500uU/ml. Sensitivity: 1.95uU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P36969; GPX4. Target Synonyms: Glutathione peroxidase 4; GPX 4; GPX-4; GPX4; GPX4_HUMAN; GSHPx-4; MCSP; mitochondrial; PHGPx; Phospholipid hydroperoxidase; Phospholipid hydroperoxide glutathione peroxidase; Phospholipid hydroperoxide glutathione peroxidase mitochondrial; snGPx; snPHGPx; Sperm nucleus glutathione peroxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4210