Learn More
Biomatik Corporation Human Pikachurin (Egflam) Elisa Kit
Supplier:  Biomatik Corporation EKC35022
Encompass_Preferred
Human Pikachurin (Egflam) Elisa Kit for detecting Egf-Like, Fibronectin Type Iii And Laminin G Domains in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q63HQ2; EGFLAM. Target Synonyms: Agrin-like protein; AGRINL; AGRNL; EGF-like; EGF-like; fibronectin type III and laminin G domains; EGF-like; fibronectin type-III and laminin G-like domain-containing protein; EGFLA_HUMAN; EGFLAM; fibronectin type-III and laminin G-like domain-containing protein; FLJ39155; Pikachurin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4212