Learn More
Biomatik Corporation Human forkhead box O3, FOXO3 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC33745
Encompass_Preferred
The Human forkhead box O3, FOXO3 ELISA Kit allows for quantitative determination of forkhead box O3 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human. Detection Range: 1.56 pg/mL-100 pg/mL. Sensitivity: 0.39 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: O43524. Restriction: For Research Use Only.
Catalog No. 50-149-7910