Learn More
Biomatik Corporation Human Platelet Receptor Gi24 (C10Orf54) Elisa Kit
Supplier:  Biomatik Corporation EKC35050
Encompass_Preferred
Human Platelet Receptor Gi24 (C10Orf54) Elisa Kit for detecting Chromosome 10 Open Reading Frame 54 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9H7M9; VSIR. Target Synonyms: B7 H5; B7H5; C10orf54; chromosome 10 open reading frame 54; DD1alpha; GI24; GI24_HUMAN; PD-1H; PDCD1 homolog; Platelet receptor Gi24; PP2135; sisp 1; SISP1; stress induced secreted protein 1; UNQ730/PRO1412; V domain Ig suppressor of T cell activation; V set domain containing immunoregulatory receptor; V set immunoregulatory receptor; V type immunoglobulin domain containing suppressor of T cell activation; VISTA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4221