Learn More
Biomatik Corporation Human Pote Ankyrin Domain Family Member E (Potee) Elisa Kit
Supplier:  Biomatik Corporation EKC35076
Encompass_Preferred
Human Pote Ankyrin Domain Family Member E (Potee) Elisa Kit for detecting Pote Ankyrin Domain Family Member E in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q6S8J3; POTEE. Target Synonyms: POTEE; A26C1A; POTE2; POTE ankyrin domain family member E; ANKRD26-like family C member 1A; Prostate; ovary; testis-expressed protein on chromosome 2; POTE-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4230