Learn More
Biomatik Corporation Human Pregnancy Specific Β1 Glycoprotein (Sp1/Psβ1-G) Elisa Kit
Supplier:  Biomatik Corporation EKC35082
Encompass_Preferred
Human Pregnancy Specific Β1 Glycoprotein (Sp1/Psβ1-G) Elisa Kit for detecting Pregnancy Specific Β1 Glycoprotein (Sp1/Psβ1-G) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.0-400ug/ml. Sensitivity: 1.0ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P11464; PSG1. Target Synonyms: B1G1; Carcinoembryonic antigen SG8; CD66 antigen-like family member F; CD66f; CD66f antigen; CEA; DHFRP2; Fetal liver non-specific cross-reactive antigen 1/2; FL-NCA-1/2; PBG 1; PBG1; Pregnancy specific B 1 glycoprotein; Pregnancy specific beta 1 glycoprotein 1; Pregnancy specific beta 1 glycoprotein 2; Pregnancy specific beta 1 glycoprotein 7; Pregnancy specific beta 1 glycoprotein 8; Pregnancy specific beta 1 glycoprotein; Pregnancy specific beta 1 glycoprotein C/D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4232