Learn More
Biomatik Corporation Human Prkc Apoptosis Wt1 Regulator Protein (Pawr) Elisa Kit
Supplier:  Biomatik Corporation EKC35093
Encompass_Preferred
Human Prkc Apoptosis Wt1 Regulator Protein (Pawr) Elisa Kit for detecting Prkc, Apoptosis, Wt1, Regulator in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q96IZ0; PAWR. Target Synonyms: 2310001G03Rik; PAR 4; PAR-4; Pawr; PAWR_HUMAN; PRKC Apoptosis WT1 Regulator; PRKC apoptosis WT1 regulator protein; Prostate apoptosis response 4 protein; Prostate apoptosis response protein 4; prostate apoptosis response protein PAR-4; Transcriptional repressor Par-4-like protein PAWR; Transcriptional repressor PAR4; WT1 Interacting Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4236