Learn More
Biomatik Corporation Human Proenkephalin (Penk) Elisa Kit
Supplier:  Biomatik Corporation EKC35116
Encompass_Preferred
Human Proenkephalin (Penk) Elisa Kit for detecting Proenkephalin in serum, plasma, cerebrospinal fluid (csf). Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01210; PENK. Target Synonyms: Leu-enkephalin; Met-enkephalin; Met-enkephalin-Arg-Gly-Leu; Met-enkephalin-Arg-Phe; Methionine enkephalin; OGF; Opioid growth factor; PENK; PENK (114-133); PENK (143-183); PENK (237-258); PENK_HUMAN; PPA; Proenkephalin A; Proenkephalin; Proenkephalin-A; Synenkephalin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4240