Learn More
Biomatik Corporation Human Prohibitin (Phb) Elisa Kit
Supplier:  Biomatik Corporation EKC35131
Encompass_Preferred
Human Prohibitin (Phb) Elisa Kit for detecting Prohibitin in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P35232; PHB. Target Synonyms: Epididymis luminal protein 215; Epididymis secretory sperm binding protein Li 54e; HEL 215; HEL S 54e; PHB; PHB_HUMAN; PHB1; Prohibitin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4243