Learn More
Biomatik Corporation Human Prostaglandin-H2 D-Isomerase (Ptgds) Elisa Kit
Supplier:  Biomatik Corporation EKC35163
Encompass_Preferred
Human Prostaglandin-H2 D-Isomerase (Ptgds) Elisa Kit for detecting Prostaglandin D2 Synthase 21Kda (Brain) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.2ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P41222; PTGDS. Target Synonyms: Beta trace protein; Beta-trace protein; Cerebrin 28; Cerebrin-28; glutathione independent PGD synthase; Glutathione independent PGD synthetase; Glutathione-independent PGD synthase; L PGDS; Lipocalin type prostaglandin D synthase; Lipocalin-type prostaglandin-D synthase; LPGDS; PDS; PGD2; PGD2 synthase; PGDS; PGDS2; prostaglandin D synthase; prostaglandin D2 synthase (21 kDa; brain); prostaglandin D2 synthase (brain); prostaglandin D2 synthase 21kDa (brain); Prostaglandin D2 synthase; prostaglandin H2 D isomerase; Prostaglandin-D2 synthase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4248