Learn More
Biomatik Corporation Human Proteasome (Prosome, Macropain) Subunit, Beta Type, 2 (Psmb2) Elisa Kit
Supplier:  Biomatik Corporation EKC35169
Encompass_Preferred
Human Proteasome (Prosome, Macropain) Subunit, Beta Type, 2 (Psmb2) Elisa Kit for detecting Proteasome (Prosome, Macropain) Subunit, Beta Type, 2 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P49721; PSMB2. Target Synonyms: HC7 I; Macropain subunit C7 I; Macropain subunit C7-I; Multicatalytic endopeptidase complex subunit C7 1; Multicatalytic endopeptidase complex subunit C7 I; Multicatalytic endopeptidase complex subunit C7-I; Proteasome (prosome; macropain) subunit beta type 2; Proteasome beta 2 subunit; Proteasome component C7 I; Proteasome component C7-I; Proteasome subunit beta type-2; PSB2_HUMAN; Psmb2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4250