Learn More
Biomatik Corporation Human Protein S Elisa Kit
Supplier:  Biomatik Corporation EKC35230
Encompass_Preferred
Human Protein S Elisa Kit for detecting Protein S in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P07225; PROS1. Target Synonyms: Preproprotein S; Propiece of latent protein S; PROS 1; PROS; PROS_HUMAN; proS1; Protein S alpha; Protein Sa; PS 21; PS 22; PS 23; PS 24; PS 25; PS 26; PS21; PS22; PS23; PS24; PS25; PS26; PSA; THPH5; THPH6; Vitamin K dependent protein S; Vitamin K-dependent plasma protein S; Vitamin K-dependent protein S. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4267