Learn More
Biomatik Corporation Human Protein-Glutamine Gamma-Glutamyltransferase 4 (Tgm4) Elisa Kit
Supplier:  Biomatik Corporation EKC35270
Encompass_Preferred
Human Protein-Glutamine Gamma-Glutamyltransferase 4 (Tgm4) Elisa Kit for detecting Transglutaminase 4 (Prostate) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 50-3200pg/ml. Sensitivity: 12.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P49221; TGM4. Target Synonyms: Dorsal prostate transglutaminase; Dorsal protein 1; Dp1; Fibrinoligase; hTGP; Prostate specific transglutaminase; Prostate transglutaminase; Prostate-specific transglutaminase; Protein glutamine gamma glutamyltransferase 4; Protein-glutamine gamma-glutamyltransferase 4; TG (P); TGase P; TGase-4; TGase4; Tgm4; TGM4_HUMAN; TGP; Transglutaminase 4 (prostate); Transglutaminase 4; Transglutaminase P; Transglutaminase; prostate; Transglutaminase-4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4272