Learn More
Biomatik Corporation Human Protocadherin-10 (Pcdh10) Elisa Kit
Supplier:  Biomatik Corporation EKC35283
Encompass_Preferred
Human Protocadherin-10 (Pcdh10) Elisa Kit for detecting Protocadherin 10 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9P2E7; PCDH10. Target Synonyms: DKFZp761O2023; KIAA1400; MGC133344; OL PCDH; Ortholog of OL pcdh; OTTHUMP00000220001; PCD10_HUMAN; PCDH10; PCDH19; Protocadherin 10; Protocadherin-10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4275