Learn More
Biomatik Corporation Human Pulmonary Activation Regulated Chemokine, Parc Elisa Kit
Supplier:  Biomatik Corporation EKC35297
Encompass_Preferred
Human Pulmonary Activation Regulated Chemokine, Parc Elisa Kit for detecting Chemokine (C-C Motif) Ligand 18 (Pulmonary And Activation-Regulated) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 4.69-300ng/ml. Sensitivity: 1.17ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P55774; CCL18. Target Synonyms: Alternative macrophage activation associated CC chemokine 1; Alternative macrophage activation-associated CC chemokine 1; AMAC-1; AMAC1; CC chemokine PARC; CCL18; CCL18 (4-69); CCL18_HUMAN; CKb7; DC-CK1; DCCK1; DCCK1; Dendritic cell chemokine 1; Macrophage inflammatory protein 4; MIP-4; MIP4; PARC; Pulmonary and activation regulated chemokine; Pulmonary and activation-regulated chemokine; SCYA18; Small inducible cytokine A18; Small inducible cytokine A18; Small-inducible cytokine A18. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4277