Learn More
Biomatik Corporation Human Relaxin, Rln Elisa Kit
Supplier:  Biomatik Corporation EKC35363
Encompass_Preferred
Human Relaxin, Rln Elisa Kit for detecting Relaxin 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P04808; RLN1. Target Synonyms: H1; H1RLX; Preprorelaxin H1; Prorelaxin; Prorelaxin H1; REL1_HUMAN; Relaxin A chain; Relaxin H1; RLN 1; Rln1; RLXH1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4282