Learn More
Biomatik Corporation Human Renin (Ren) Elisa Kit
Supplier:  Biomatik Corporation EKC35366
Encompass_Preferred
Human Renin (Ren) Elisa Kit for detecting Renin in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000mIU/ml. Sensitivity: 3.9mIU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P00797; REN. Target Synonyms: Angiotensin forming enzyme; Angiotensin forming enzyme precursor; Angiotensinogenase; Angiotensinogenase precursor; FLJ10761; HNFJ2; REN; Ren1; RENI_HUMAN; Renin; Renin precursor renal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4284