Learn More
Biomatik Corporation Human Reticulon-4 Receptor-Like 2 (Rtn4Rl2) Elisa Kit
Supplier:  Biomatik Corporation EKC35376
Encompass_Preferred
Human Reticulon-4 Receptor-Like 2 (Rtn4Rl2) Elisa Kit for detecting Reticulon 4 Receptor-Like 2 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q86UN3; RTN4RL2. Target Synonyms: RTN4RL2; NGRH1; NGRL3; Reticulon-4 receptor-like 2; Nogo receptor-like 3; Nogo-66 receptor homolog 1; Nogo-66 receptor-related protein 2; NgR2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4286