Learn More
Biomatik Corporation Human Soluble CD276,sCD276/sB7-H3 ELISA Kit
Supplier:  Biomatik Corporation EKC35590
Encompass_Preferred
Sample Type: serum, plasma, urine, cell culture supernates. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12 ng/mL-200 ng/mL. Sensitivity: 0.78 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q5ZPR3. Target Synonyms: B7-H3, B7H3, B7 homolog 3|CD276 antigen. Restriction: For Research Use Only.
Catalog No. 50-229-4302