Learn More
Biomatik Corporation Human Soluble Platelet Endothelial Cell Adhesion Molecule 1, Specam-1 Elisa Kit
Supplier:  Biomatik Corporation EKC35615
Encompass_Preferred
Human Soluble Platelet Endothelial Cell Adhesion Molecule 1, Specam-1 Elisa Kit for detecting Soluble Platelet Endothelial Cell Adhesion Molecule 1, Specam-1 in serum, plasma, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P16284; PECAM1. Target Synonyms: PECAM1; Platelet endothelial cell adhesion molecule; PECAM-1; EndoCAM; GPIIA'; PECA1; CD antigen CD31. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4305