Learn More
Biomatik Corporation Human Tetranectin (Clec3B) Elisa Kit
Supplier:  Biomatik Corporation EKC35749
Encompass_Preferred
Human Tetranectin (Clec3B) Elisa Kit for detecting Tetranectin(Clec3B) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P05452; CLEC3B. Target Synonyms: C type lectin domain family 3 member B; C-type lectin domain family 3 member B; Clec3b; Plasminogen kringle 4 binding protein; Plasminogen kringle 4-binding protein; TETN_HUMAN; Tetranectin (plasminogen binding protein); Tetranectin; TN; TNA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4315