Learn More
Biomatik Corporation Human Thymidinephosphorylase, Tp Elisa Kit
Supplier:  Biomatik Corporation EKC35779
Encompass_Preferred
Human Thymidinephosphorylase, Tp Elisa Kit for detecting Thymidine Phosphorylase in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000ng/ml. Sensitivity: 15.6ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P19971; TYMP. Target Synonyms: ECGF 1; ECGF; ECGF1; Endothelial cell growth factor 1; Endothelial cell growth factor 1 platelet derived; Endothelial cell growth factor; platelet-derived; Gliostatin; hPD ECGF; MEDPS1; MNGIE; MTDPS1; PD ECGF; PD-ECGF; PDECGF; PDEGF; Platelet derived endothelial cell growth factor; Platelet derived endothelial growth factor; Platelet-derived endothelial cell growth factor; TdRPase; Thymidine phosphorylase; TP; Tymp; TYPH_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4320