Learn More
Biomatik Corporation Human Transitional Endoplasmic Reticulum Atpase (Vcp) Elisa Kit
Supplier:  Biomatik Corporation EKC35861
Encompass_Preferred
Human Transitional Endoplasmic Reticulum Atpase (Vcp) Elisa Kit for detecting Valosin-Containing Protein in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.44-1500pg/ml. Sensitivity: 5.86pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P55072; VCP. Target Synonyms: 15S Mg (2+) ATPase p97 subunit; 15S Mg (2+)-ATPase p97 subunit; ALS14; ATPase p97; CDC48; IBMPFD; MGC131997; MGC148092; MGC8560; p97; TER ATPase; TERA; TERA_HUMAN; Transitional endoplasmic reticulum ATPase; Valosin containing protein; Valosin-containing protein; VCP; Yeast Cdc48p homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4324