Learn More
Biomatik Corporation Human Trem-Like Transcript 1 Protein (Treml1) Elisa Kit
Supplier:  Biomatik Corporation EKC35874
Encompass_Preferred
Human Trem-Like Transcript 1 Protein (Treml1) Elisa Kit for detecting Triggering Receptor Expressed On Myeloid Cells-Like 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q86YW5; TREML1. Target Synonyms: dJ238O23.3; GLTL1825; MGC119173; PRO3438; TLT 1; TLT-1; TLT1; TREM like transcript 1; Trem like transcript 1 protein; Trem-like transcript 1 protein; TREML 1; Treml1; Triggering receptor expressed on myeloid cells like 1; Triggering receptor expressed on myeloid cells like protein 1; Triggering receptor expressed on myeloid cells-like protein 1; TRML1_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4326