Learn More
Biomatik Corporation Monkey Somatotropin(GH1) ELISA kit
Supplier:  Biomatik Corporation EKC36199
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Monkey. Detection Range: 2.5 ng/mL-50 ng/mL. Sensitivity: 1 ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P33093. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4355