Learn More
Biomatik Corporation Monkey Vitamin K-dependent protein S(PROS1) ELISA kit
Supplier:  Biomatik Corporation EKC36205
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Monkey. Detection Range: 15.6 ng/mL-1000 ng/mL. Sensitivity: 3.9 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q28520. Target Synonyms: PROS, PS21, PS22, PS23, PS24, PS25, PSA, protein S, alpha|protein Sa|vitamin K-dependent plasma protein S. Restriction: For Research Use Only.
Catalog No. 50-229-4357