Learn More
Biomatik Corporation Mouse 6-keto-prostaglandin F1a,6-K-PGF1a ELISA Kit
Supplier:  Biomatik Corporation EKC36218
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 25 pg/mL-1000 pg/mL. Sensitivity: 25 pg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4359