Learn More
Biomatik Corporation Human gamma glutamyl transpeptidase, GGT ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC33784
Encompass_Preferred
The Human gamma glutamyl transpeptidase, GGT ELISA Kit allows for quantitative determination of gamma-glutamyltransferase 1 in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 0.78 mU/mL-50 mU/mL. Sensitivity: 0.195mU/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P19440. Restriction: For Research Use Only.
Catalog No. 50-149-7915