Learn More
Biomatik Corporation Mouse B-cell leukemia/lymphoma 2,Bcl-2 ELISA Kit
Supplier:  Biomatik Corporation EKC36393
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.6 pg/mL-1000 pg/mL. Sensitivity: 3.9 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P10417. Target Synonyms: Bcl-2, B-cell lymphoma protein 2|OTTHUMP00000163680. Restriction: For Research Use Only.
Catalog No. 50-229-4369