Learn More
Biomatik Corporation Mouse Bone Morphogenetic Protein 9, Bmp-9 Elisa Kit
Supplier:  Biomatik Corporation EKC36426
Encompass_Preferred
Mouse Bone Morphogenetic Protein 9, Bmp-9 Elisa Kit for detecting Growth Differentiation Factor 2 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20pg/ml. Sensitivity: 0.078pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9WV56; Gdf2. Target Synonyms: Gdf2; Bmp9Growth/differentiation factor 2; GDF-2; Bone morphogenetic protein 9; BMP-9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4373