Learn More
Biomatik Corporation Mouse Chordin-Like Protein 1 (Chrdl1) Elisa Kit
Supplier:  Biomatik Corporation EKC36551
Encompass_Preferred
Mouse Chordin-Like Protein 1 (Chrdl1) Elisa Kit for detecting Chordin-Like 1 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q920C1; Chrdl1. Target Synonyms: Chrdl1; Ng1; Nrln1Chordin-like protein 1; Neuralin-1; Neurogenesin-1; Ventroptin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4382