Learn More
Biomatik Corporation Mouse coagulation factor Ⅺ,FⅪ ELISA Kit
Supplier:  Biomatik Corporation EKC36576
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.47 ng/mL-30 ng/mL. Sensitivity: 0.12 ng/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q91Y47. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4384